Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CITED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168388.2 | NP_001161860.1 | 2 | 1033 | ||
NM_001168389.2 | NP_001161861.2 | 2 | 1016 | |||
NM_006079.4 | NP_006070.2 | 2 | 1172 | |||
GenBank mRNA | AF109161.1 | - | 2 | 1142 | ||
BC004377.1 | - | 2 | 1125 | 3640855 |