Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009873.2 | NP_034003.1 | 2, 3 | 432 | ||
GenBank mRNA | AF132482.1 | - | 2, 3 | 263 | ||
AF132483.1 | - | 2, 3 | 432 | |||
AK078973.1 | - | 2, 3 | 543 | |||
AK154197.1 | - | 3, 4 | 705 | |||
BC119360.1 | - | 2, 3 | 421 | 8734109 | ||
BC120595.1 | - | 2, 3 | 421 | 40129518 |