Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009892.2 | NP_034022.2 | 601 | |||
GenBank mRNA | AK137503.1 | - | 578 | |||
AK154420.1 | - | 576 | ||||
BC061154.1 | - | 576 | 30298306 | |||
D87757.2 | - | 6 | 558 | |||
M94584.2 | - | 570 | ||||
U56900.1 | - | 2 | 145 |