Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111021.2 | NP_001104491.1 | 8 | 2803 | ||
NM_001111022.2 | NP_001104492.1 | 7 | 2611 | |||
NM_001111023.2 | NP_001104493.1 | 6 | 4138 | |||
NM_009821.3 | NP_033951.2 | 5 | 3946 | |||
GenBank mRNA | AK051758.1 | - | 5 | 3484 | ||
AK137493.1 | - | 8 | 2773 | |||
AK145098.1 | - | 8 | 2803 | |||
AK153274.1 | - | 1 | 1188 | |||
AK165307.1 | - | 8 | 2826 | |||
D26532.1 | - | 5 | 3950 |