Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111026.2 | NP_001104496.1 | 11 | 2801 | ||
NM_001111027.2 | NP_001104497.1 | 12 | 2995 | |||
NM_009822.3 | NP_033952.1 | 11 | 2733 | |||
GenBank mRNA | AK032132.1 | - | 11 | 2668 | ||
AK142006.1 | - | 11 | 2802 | |||
D32007.1 | - | 11 | 2732 |