Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfa2t3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109873.1 | NP_001103343.1 | 12 | 2655 | ||
NM_009824.2 | NP_033954.2 | 12 | 2636 | |||
NM_177289.1 | NP_796263.1 | 11 | 2577 | |||
GenBank mRNA | AF038029.1 | - | 12 | 2518 | ||
AK033680.1 | - | 11 | 2473 | |||
AK041887.1 | - | 12 | 2658 | |||
BC055951.1 | - | 4 | 1220 |