Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009831.2 | NP_033961.1 | 5 | 958 | ||
GenBank mRNA | AK128957.1 | - | 5 | 946 | ||
AK136645.1 | - | 5 | 695 | |||
AK143114.1 | - | 5 | 949 | |||
AK147026.1 | - | 5 | 946 | |||
AK153392.1 | - | 5 | 890 | |||
AK159429.1 | - | 5 | 888 | |||
AK159831.1 | - | 5 | 893 | |||
AK160411.1 | - | 5 | 933 | |||
AK167052.1 | - | 5 | 890 | |||
AK167486.1 | - | 5 | 885 | |||
AK170397.1 | - | 5 | 950 | |||
BC005534.1 | - | 4 | 608 | |||
BC006609.1 | - | 5 | 841 | |||
L49507.1 | - | 5 | 901 | |||
Z37110.1 | - | 5 | 901 |