Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271566.1 | NP_001258495.1 | 12 | 2240 | ||
NM_001271567.1 | NP_001258496.1 | 11 | 1975 | |||
NM_001271568.1 | NP_001258497.1 | 11 | 1954 | |||
NM_009863.3 | NP_033993.2 | 12 | 2071 | |||
GenBank mRNA | AB018574.1 | - | 11 | 1976 | ||
AB018575.1 | - | 12 | 2072 | |||
AJ007661.1 | - | 12 | 2023 | |||
AK145921.1 | - | 12 | 1997 | |||
AK146009.1 | - | 12 | 2241 | |||
BC062149.1 | - | 11 | 2789 | |||
BC072666.1 | - | 12 | 2012 | 30611704 | ||
BC080702.1 | - | 11 | 1907 | 30474846 |