Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024806.2 | NP_001019977.1 | 6, 7 | 2259 | ||
GenBank mRNA | AK136173.1 | - | 6, 7 | 2240 | ||
AK145374.1 | - | 6, 7 | 2241 | |||
AK155990.1 | - | 6, 7 | 2242 | |||
AK158771.1 | - | 6, 7 | 2236 | |||
AK159081.1 | - | 6, 7 | 2256 | |||
U19891.1 | - | 6, 7 | 2230 | |||
U19892.1 | - | 6, 7 | 1944 |