Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278041.1 | NP_001264970.1 | 18, 19 | 2426 | ||
NM_009893.2 | NP_034023.1 | 18, 19 | 2360 | |||
GenBank mRNA | AF069501.1 | - | 18, 19 | 2360 | ||
AK077460.1 | - | 19, 20 | 2521 | |||
AK149445.1 | - | 18, 19 | 2335 | |||
AK220310.1 | - | 2, 3 | 2721 | |||
BC145017.1 | - | 16, 17 | 2474 | |||
BC145018.1 | - | 18, 19 | 2394 | |||
BC145019.1 | - | 18, 19 | 2431 | |||
BC145696.1 | - | 18, 19 | 2424 | 40130999 |