Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042634.2 | NP_001036099.1 | 13 | 1653 | ||
GenBank mRNA | AA267385.1 | - | 2 | 231 | ||
AK076199.1 | - | 13 | 1588 | |||
AK135765.1 | - | 13 | 1588 | |||
AK146091.1 | - | 7 | 1943 | |||
BC062887.1 | - | 2863 | ||||
L29221.1 | - | 12 | 1478 | |||
M38381.1 | - | 13 | 1569 | |||
X57186.1 | - | 13 | 1553 |