Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009926.1 | NP_034056.1 | 2, 3 | 458 | ||
GenBank mRNA | D38412.1 | - | 2, 3 | 422 | ||
KJ650220.1 | - | 2, 3 | 240 | |||
U16789.1 | - | 2, 3 | 459 |