Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col9a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009936.2 | NP_034066.2 | 2, 3 | 169 | ||
GenBank mRNA | AF349718.1 | - | 1, 2 | 79 | ||
AK030350.1 | - | 2, 3 | 148 | |||
CK623954.1 | - | 2, 3 | 169 |