Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010014.3 | NP_034144.1 | 4 | 557 | ||
NM_177259.4 | NP_796233.2 | 4 | 557 | |||
GenBank mRNA | AK005640.1 | - | 3 | 368 | ||
BC051993.1 | - | 6 | 690 | |||
BC138538.1 | - | 3 | 278 | 8861558 | ||
BC138539.1 | - | 3 | 278 | 8861559 | ||
BY731247.1 | - | 4 | 524 | |||
Y08379.1 | - | 4 | 505 | |||
Y08380.1 | - | 4 | 505 | |||
Y08381.1 | - | 4 | 505 |