Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286396.1 | NP_001273325.1 | 6 | 546 | ||
NM_001286397.1 | NP_001273326.1 | 6 | 556 | |||
NM_010018.3 | NP_034148.2 | 6 | 705 | |||
GenBank mRNA | AK134813.1 | - | 6 | 566 | ||
BC015269.1 | - | 6 | 556 | 4239778 | ||
BC018377.1 | - | 6 | 546 | 4242375 | ||
D10210.1 | - | 5 | 497 | |||
D10211.1 | - | 5 | 497 | |||
M32299.1 | - | 6 | 562 |