Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009919.2 | NP_034049.2 | 4 | 453 | ||
GenBank mRNA | AF022811.1 | - | 4 | 444 | ||
AK002293.1 | - | 4 | 421 | |||
AK011200.1 | - | 4 | 406 | |||
AK152432.1 | - | 4 | 404 | |||
AK154479.1 | - | 4 | 404 | |||
AK168154.1 | - | 4 | 404 | |||
BC034868.1 | - | 4 | 413 | 2938052 | ||
DV653213.1 | - | 4 | 507 |