Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB006191.2 | - | 5 | 565 | ||
AK141483.1 | - | 5 | 665 | |||
BC059922.1 | - | 5 | 651 | |||
BC103797.1 | - | 5 | 639 | 30134114 | ||
BC137585.1 | - | 5 | 526 | 8860604 | ||
BC137586.1 | - | 5 | 526 | 8860605 | ||
BC144739.1 | - | 5 | 552 |