Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009922.4 | NP_034052.3 | 3 | 333 | ||
GenBank mRNA | AK035548.1 | - | 3 | 306 | ||
BC138863.1 | - | 3 | 265 | 8861885 | ||
BC138864.1 | - | 3 | 265 | 8861886 | ||
BC145446.1 | - | 3 | 265 | |||
Z19542.1 | - | 3 | 327 |