Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009938.4 | NP_034068.3 | 7 | 892 | ||
GenBank mRNA | AK043479.1 | - | 7 | 618 | ||
AK186354.1 | - | 1 | 34 | |||
BC024070.1 | - | 7 | 545 | 5097228 | ||
BC047429.1 | - | 7 | 583 | 3498656 | ||
BC082785.1 | - | 3 | 232 |