Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136057.2 | NP_001129529.1 | 5 | 732 | ||
NM_001146183.1 | NP_001139655.1 | 5 | 709 | |||
NM_009947.3 | NP_034077.1 | 5 | 535 | |||
GenBank mRNA | AB008893.1 | - | 5 | 535 | ||
AK134548.1 | - | 5 | 554 | |||
AK144410.1 | - | 5 | 561 | |||
AK158638.1 | - | 4 | 1954 | |||
BC050766.1 | - | 5 | 557 | 6591063 | ||
BC051994.1 | - | 4 | 1117 |