Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009949.2 | NP_034079.2 | 2, 3 | 364 | ||
GenBank mRNA | AK018752.1 | - | 2, 3 | 313 | ||
AK144441.1 | - | 2, 3 | 364 | |||
AK150163.1 | - | 2, 3 | 336 | |||
AK169038.1 | - | 2, 3 | 261 | |||
BC138514.1 | - | 2, 3 | 299 | 8861534 | ||
BC145859.1 | - | 2, 3 | 299 | 40131162 | ||
CD545813.1 | - | 2, 3 | 339 | |||
U01163.1 | - | 2, 3 | 309 |