Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009982.4 | NP_034112.3 | 2 | 389 | ||
GenBank mRNA | AK002454.1 | - | 2 | 348 | ||
AK049406.1 | - | 2 | 348 | |||
AK146858.1 | - | 2 | 397 | |||
AK150761.1 | - | 2 | 349 | |||
AK151388.1 | - | 2 | 348 | |||
AK151711.1 | - | 2 | 349 | |||
AK151861.1 | - | 2 | 235 | |||
AK152524.1 | - | 2 | 346 | |||
AK153168.1 | - | 2 | 349 | |||
AK167882.1 | - | 2 | 348 | |||
BC067063.1 | - | 2 | 322 | 30541061 | ||
U74683.3 | - | 2 | 346 | |||
U89269.1 | - | 2 | 347 |