Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRK4 SIRNA Assays ›
Gene Symbol:
GRK4
Gene Name:
G protein-coupled receptor kinase 4
Gene Aliases:
GPRK2L, GPRK4, GRK4a, IT11
Chromosome Location:
Chr.4: 2963504 - 3040760 on Build GRCh38
UniGene ID:
Hs.32959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
664
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004056.1
NP_001004056.1
8
1281
NM_001004057.1
NP_001004057.1
9
1377
NM_005307.2
NP_005298.2
8
1281
NM_182982.2
NP_892027.2
9
1377
XM_005247962.3
XP_005248019.1
7
895
XM_006713880.3
XP_006713943.1
8
793
XM_011513447.2
XP_011511749.1
10
1606
XM_011513448.2
XP_011511750.1
10
1606
XM_011513449.2
XP_011511751.1
9
1510
XM_011513450.2
XP_011511752.1
10
1606
XM_011513451.2
XP_011511753.1
10
1606
XM_011513452.2
XP_011511754.1
10
1606
XM_011513453.2
XP_011511755.1
10
1606
XM_011513454.2
XP_011511756.1
10
1606
XM_011513455.2
XP_011511757.1
10
1606
XM_011513456.2
XP_011511758.1
10
1606
XM_017008052.1
XP_016863541.1
9
1528
XM_017008053.1
XP_016863542.1
10
1606
XM_017008054.1
XP_016863543.1
9
1489
XM_017008055.1
XP_016863544.1
8
1393
XM_017008056.1
XP_016863545.1
9
1489
XM_017008057.1
XP_016863546.1
6
693
XM_017008058.1
XP_016863547.1
8
753
XM_017008059.1
XP_016863548.1
6
777
XM_017008060.1
XP_016863549.1
7
871
XM_017008061.1
XP_016863550.1
8
1144
XM_017008062.1
XP_016863551.1
8
1291
XM_017008063.1
XP_016863552.1
7
1080
XM_017008064.1
XP_016863553.1
7
864
XM_017008066.1
XP_016863555.1
6
701
GenBank mRNA
AK223581.1
-
8
1289
AK292390.1
-
9
1422
BC117320.1
-
9
1265
40125871
U33054.1
-
9
1176
U33055.1
-
8
1080
U33056.1
-
9
1176
X97879.1
-
8
924
X97880.1
-
8
924
X97881.1
-
9
1020
X98118.1
-
9
1020
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_022928, XM_008770355, XM_008770354, XM_008770357, XM_008770353, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor internalization
protein phosphorylation
signal transduction
regulation of G-protein coupled receptor protein signaling pathway
regulation of rhodopsin mediated signaling pathway
receptor internalization
G-protein coupled receptor kinase activity
ATP binding
rhodopsin kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-99mnp:80/10.239.163.72:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0