Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039162.2 | NP_001034251.1 | 5 | 1319 | ||
NM_009990.3 | NP_034120.2 | 5 | 1319 | |||
GenBank mRNA | AK045967.1 | - | 5 | 1163 | ||
AK172928.1 | - | 5 | 1212 | |||
BC039162.1 | - | 5 | 1259 | 3498067 | ||
BC053048.1 | - | 5 | 1319 | 5709279 | ||
BC121191.1 | - | 5 | 965 |