Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010008.4 | NP_034138.3 | 3, 4 | 727 | ||
GenBank mRNA | AK033614.1 | - | 3, 4 | 730 | ||
AK045421.1 | - | 3, 4 | 717 | |||
AK078893.1 | - | 3, 4 | 553 | |||
AK084025.1 | - | 3, 4 | 737 | |||
AK165663.1 | - | 3, 4 | 572 | |||
BC050832.1 | - | 3, 4 | 721 | 6514145 | ||
U62295.1 | - | 3, 4 | 774 |