Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002848.3 | NP_002839.1 | 18, 19 | 3223 | ||
NM_030667.2 | NP_109592.1 | 19, 20 | 3307 | |||
NM_030668.2 | NP_109593.1 | 6, 7 | 814 | |||
NM_030669.2 | NP_109594.1 | 7, 8 | 898 | |||
NM_030670.2 | NP_109595.1 | 6, 7 | 814 | |||
NM_030671.2 | NP_109596.1 | 7, 8 | 898 | |||
GenBank mRNA | AF152378.1 | - | 7, 8 | 890 | ||
AF187042.1 | - | 6, 7 | 806 | |||
AF187043.1 | - | 7, 8 | 890 | |||
AF187044.1 | - | 6, 7 | 806 | |||
AK289784.1 | - | 19, 20 | 3052 | |||
AK290982.1 | - | 6, 7 | 537 | |||
AK299529.1 | - | 4, 5 | 273 | |||
BC126201.1 | - | 19, 20 | 3007 | 8991917 | ||
BC126203.1 | - | 19, 20 | 3007 | 8991919 | ||
U20489.1 | - | 18, 19 | 2925 | |||
Z48541.1 | - | 19, 20 | 2957 |