Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009908.2 | NP_034038.2 | 6 | 972 | ||
GenBank mRNA | AJ006215.1 | - | 6 | 885 | ||
AK087150.1 | - | 3 | 274 | |||
AK155730.1 | - | 6 | 925 | |||
AK218506.1 | - | 2 | 145 | |||
BC004606.1 | - | 1 | 89 | |||
BC031500.1 | - | 6 | 901 | 2647633 | ||
BC063776.1 | - | 6 | 901 | 6398450 |