Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009917.5 | NP_034047.2 | 2 | 2145 | ||
GenBank mRNA | AK154595.1 | - | 2 | 2146 | ||
AK155628.1 | - | 2 | 2119 | |||
AK155632.1 | - | 2 | 2117 | |||
AK171391.1 | - | 604 | ||||
D83648.1 | - | 2 | 2111 |