Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB006191.2 | - | 6 | 839 | ||
AK141483.1 | - | 6 | 939 | |||
BC059922.1 | - | 6 | 925 | |||
BC103797.1 | - | 6 | 913 | 30134114 | ||
BC137585.1 | - | 6 | 800 | 8860604 | ||
BC137586.1 | - | 6 | 800 | 8860605 | ||
BC144739.1 | - | 6 | 826 | |||
CX243776.1 | - | 1 | 207 |