Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146318.1 | NP_001139790.1 | 3 | 2002 | ||
NM_009923.2 | NP_034053.2 | 4 | 1787 | |||
GenBank mRNA | AK041969.1 | - | 3 | 2002 | ||
AK087415.1 | - | 4 | 1754 | |||
AK140280.1 | - | 3 | 2003 | |||
AK154654.1 | - | 4 | 1809 | |||
AK158318.1 | - | 4 | 1782 | |||
AK158520.1 | - | 4 | 1704 | |||
AK172318.1 | - | 4 | 1750 | |||
AK172514.1 | - | 4 | 1750 | |||
AK205418.1 | - | 1 | 203 | |||
BC005544.1 | - | 4 | 1741 | 3585034 | ||
BC021904.1 | - | 4 | 1677 | 3669076 | ||
M31810.1 | - | 3 | 2368 |