Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009950.2 | NP_034080.1 | 2 | 227 | ||
GenBank mRNA | AF017992.1 | - | 1 | 108 | ||
AJ224738.1 | - | 2 | 226 | |||
AJ224740.1 | - | 2 | 212 | |||
AK052304.1 | - | 2 | 132 | |||
AK131713.1 | - | 2 | 334 | |||
AK136561.1 | - | 2 | 132 | |||
AK144648.1 | - | 2 | 248 | |||
AK163112.1 | - | 2 | 239 | |||
BC005608.1 | - | 1 | 190 | 3498522 |