Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009969.4 | NP_034099.2 | 4 | 814 | ||
GenBank mRNA | AK053196.1 | - | 4 | 814 | ||
BC116878.1 | - | 4 | 548 | 40126197 | ||
BC116880.1 | - | 4 | 548 | 40126199 | ||
X02333.1 | - | 4 | 698 | |||
X03019.1 | - | 4 | 557 | |||
X03221.1 | - | 4 | 556 | |||
X05906.1 | - | 4 | 456 |