Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170744.1 | NP_001164215.1 | 6 | 2470 | ||
NM_009980.4 | NP_034110.1 | 8 | 1347 | |||
GenBank mRNA | AB033123.1 | - | 8 | 1212 | ||
AF016508.1 | - | 6 | 819 | |||
AF059735.1 | - | 6 | 798 | |||
AK134399.1 | - | 7 | 871 | |||
AK146713.1 | - | 8 | 1232 | |||
AK147871.1 | - | 8 | 1230 | |||
AK217631.1 | - | 3 | 257 | |||
BC013333.1 | - | 6 | 854 | 4019572 | ||
BC017520.1 | - | 7 | 2315 | 4511826 |