Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009989.3 | NP_034119.1 | 3 | 338 | ||
GenBank mRNA | AK005582.1 | - | 3 | 272 | ||
AK018833.1 | - | 3 | 272 | |||
AK018851.1 | - | 3 | 272 | |||
BC049608.1 | - | 3 | 292 | |||
BC059728.1 | - | 3 | 334 | 6741773 | ||
M20625.1 | - | 3 | 333 | |||
X55771.1 | - | 3 | 271 |