Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010027.1 | NP_034157.1 | 3 | 393 | ||
GenBank mRNA | AK144193.1 | - | 3 | 436 | ||
BC010753.1 | - | 3 | 387 | 4241633 |