Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319210.1 | NP_001306139.1 | 2 | 570 | ||
NM_001319211.1 | NP_001306140.1 | 2 | 202 | |||
NM_001319212.1 | NP_001306141.1 | 2 | 570 | |||
NM_003672.3 | NP_003663.2 | 2 | 570 | |||
NM_033312.2 | NP_201569.1 | 2 | 570 | |||
NM_033313.2 | NP_201570.1 | 2 | 570 | |||
XM_005271294.3 | XP_005271351.1 | 2 | 110 | |||
XM_005271296.3 | XP_005271353.1 | 2 | 107 | |||
XM_011542340.2 | XP_011540642.1 | 2 | 109 | |||
XM_011542346.2 | XP_011540648.1 | 2 | 118 | |||
XM_017002646.1 | XP_016858135.1 | 2 | 110 | |||
XM_017002647.1 | XP_016858136.1 | 2 | 651 | |||
GenBank mRNA | AF000367.1 | - | 2 | 437 | ||
AF064102.1 | - | 2 | 545 | |||
AF064103.1 | - | 2 | 535 | |||
AF122013.1 | - | 2 | 82 | |||
AK302310.1 | - | 2 | 191 | |||
AK310657.1 | - | 2 | 528 | |||
BC038979.1 | - | 2 | 144 | |||
BC093916.1 | - | 2 | 177 | 7939761 | ||
BC093918.1 | - | 2 | 177 | 7939763 | ||
DA885382.1 | - | 2 | 399 | |||
DB029799.1 | - | 2 | 215 | |||
DQ530256.1 | - | 2 | 88 |