Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190703.1 | NP_001177632.1 | 3 | 420 | ||
NM_001190704.1 | NP_001177633.1 | 3 | 420 | |||
NM_001190705.1 | NP_001177634.1 | 3 | 420 | |||
NM_010052.5 | NP_034182.2 | 3 | 420 | |||
GenBank mRNA | AK005522.1 | - | 3 | 385 | ||
AK133664.1 | - | 4 | 361 | |||
AK162458.1 | - | 3 | 421 | |||
AK164935.1 | - | 3 | 385 | |||
BC052159.1 | - | 3 | 368 | 6581768 | ||
EU434914.1 | - | 2 | 149 | |||
EU434915.1 | - | 3 | 220 | |||
EU434916.1 | - | 2 | 113 | |||
EU434917.1 | - | 3 | 340 | |||
L12721.1 | - | 3 | 368 | |||
U15980.1 | - | 3 | 348 | |||
Z12171.1 | - | 3 | 215 |