Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010071.2 | NP_034201.1 | 3 | 476 | ||
GenBank mRNA | AF030627.1 | - | 3 | 476 | ||
AF035117.1 | - | 3 | 472 | |||
AF059583.1 | - | 3 | 462 | |||
AK155579.1 | - | 3 | 473 | |||
AK158246.1 | - | 3 | 440 | |||
AK159466.1 | - | 3 | 517 | |||
AK159472.1 | - | 3 | 554 | |||
BC004590.1 | - | 3 | 468 | 3587682 |