Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033343.3 | NP_203129.1 | 2, 3 | 474 | ||
XM_011510105.2 | XP_011508407.1 | 2, 3 | 708 | |||
XM_011510106.2 | XP_011508408.1 | 2, 3 | 331 | |||
GenBank mRNA | AB055703.1 | - | 1, 2 | 177 | ||
AF179849.1 | - | 2, 3 | 478 | |||
AF282899.1 | - | 1, 2 | 173 | |||
AY053457.1 | - | 2, 3 | 266 | |||
BC011759.2 | - | 2, 3 | 285 | 3529089 | ||
KU178818.1 | - | 2, 3 | 242 |