Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010112.4 | NP_034242.2 | 2 | 1506 | ||
GenBank mRNA | AK030321.1 | - | 2 | 633 | ||
AK163191.1 | - | 2 | 547 | |||
BC005438.1 | - | 2 | 575 | 3487600 | ||
BC031543.1 | - | 2 | 301 | 3669513 | ||
U28728.1 | - | 2 | 530 | |||
U57686.1 | - | 2 | 230 |