Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010120.5 | NP_034250.3 | 3 | 460 | ||
GenBank mRNA | AF026481.1 | - | 2 | 1279 | ||
AK009026.1 | - | 3 | 431 | |||
AK013268.1 | - | 3 | 459 | |||
AK019451.1 | - | 3 | 461 | |||
AK150651.1 | - | 3 | 457 | |||
AK150673.1 | - | 3 | 433 | |||
AK150677.1 | - | 3 | 433 | |||
AK166368.1 | - | 432 | ||||
AK167333.1 | - | 3 | 420 | |||
BC027437.1 | - | 3 | 422 | 4017054 | ||
CT010335.1 | - | 1 | 107 |