Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010151.2 | NP_034281.2 | 1 | 755 | ||
GenBank mRNA | BC048695.1 | - | 1 | 441 | ||
BC108408.1 | - | 1 | 644 | 30245025 | ||
U07625.1 | - | 1 | 357 | |||
X74134.1 | - | 1 | 655 |