Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010157.3 | NP_034287.3 | 3 | 914 | ||
NM_207707.1 | NP_997590.1 | 3 | 914 | |||
GenBank mRNA | AK054290.1 | - | 3 | 912 | ||
AK054301.1 | - | 3 | 915 | |||
AK054413.1 | - | 3 | 915 | |||
AY054413.1 | - | 2 | 375 | |||
BC141075.1 | - | 2 | 616 | 9055687 | ||
BC145329.1 | - | 2 | 616 | 9053885 | ||
U81451.1 | - | 2 | 386 |