Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010049.3 | NP_034179.1 | 2, 3 | 620 | ||
GenBank mRNA | AK031241.1 | - | 2, 3 | 254 | ||
AK136212.1 | - | 2, 3 | 678 | |||
BC005796.1 | - | 2, 3 | 192 | 3707582 | ||
BU058996.1 | - | 2, 3 | 620 | |||
L26316.1 | - | 2, 3 | 199 | |||
V00734.1 | - | 2, 3 | 256 | |||
X56066.1 | - | 2, 3 | 166 |