Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010053.1 | NP_034183.1 | 2 | 930 | ||
GenBank mRNA | AB221644.1 | - | 2 | 381 | ||
BC058394.1 | - | 2 | 428 | |||
BC079609.1 | - | 2 | 788 | 30360273 | ||
U51001.1 | - | 2 | 930 |