Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010072.3 | NP_034202.1 | 6 | 445 | ||
GenBank mRNA | AB004789.1 | - | 6 | 419 | ||
AI099209.1 | - | 6 | 445 | |||
AK004834.1 | - | 6 | 438 | |||
AK005268.1 | - | 6 | 431 | |||
AK008548.1 | - | 6 | 433 | |||
AK147481.1 | - | 1 | 2473 | |||
BC061151.1 | - | 6 | 425 | 30307514 |