Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159543.1 | NP_001153015.1 | 9 | 1124 | ||
NM_010074.3 | NP_034204.1 | 9 | 1124 | |||
GenBank mRNA | AK040409.1 | - | 8 | 830 | ||
AK085370.1 | - | 9 | 707 | |||
AK153939.1 | - | 9 | 738 | |||
AK163085.1 | - | 9 | 1129 | |||
AK164775.1 | - | 9 | 707 | |||
BC022183.1 | - | 9 | 986 | 3982742 | ||
X58384.1 | - | 9 | 801 |