Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285807.1 | NP_001272736.1 | 7 | 999 | ||
NM_001285808.1 | NP_001272737.1 | 5 | 655 | |||
NM_001285810.1 | NP_001272739.1 | 5 | 655 | |||
NM_001285811.1 | NP_001272740.1 | 6 | 777 | |||
NM_001285813.1 | NP_001272742.1 | 6 | 812 | |||
NM_001285817.1 | NP_001272746.1 | 5 | 655 | |||
NM_010087.4 | NP_034217.2 | 6 | 777 | |||
NM_207650.4 | NP_997533.1 | 6 | 777 | |||
GenBank mRNA | AF143542.1 | - | 6 | 676 | ||
AF143543.1 | - | 6 | 713 | |||
AF143544.1 | - | 6 | 622 | |||
AK003829.2 | - | 6 | 697 | |||
AK019477.1 | - | 7 | 919 | |||
AK131649.1 | - | 5 | 548 | |||
AK147279.1 | - | 6 | 723 | |||
AK147359.1 | - | 5 | 612 | |||
BC040364.1 | - | 6 | 744 | 4505583 | ||
BC050765.1 | - | 5 | 563 | 6533769 | ||
X95226.1 | - | 5 | 477 | |||
X95227.1 | - | 6 | 607 |