Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037957.3 | NP_001033046.1 | 4 | 498 | ||
NM_001271370.1 | NP_001258299.1 | 4 | 451 | |||
NM_010092.2 | NP_034222.1 | 4 | 498 | |||
GenBank mRNA | AJ537610.1 | - | 4 | 451 | ||
AK148342.1 | - | 4 | 478 | |||
BC019545.1 | - | 4 | 498 | 4511845 | ||
Y18280.2 | - | 4 | 487 |