Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034996.1 | - | 3, 4 | 1254 | ||
BC006797.1 | - | 3, 4 | 1223 | 2648527 | ||
BC018572.1 | - | 3, 4 | 1064 | |||
BC021656.1 | - | 3, 4 | 1236 | 3602195 | ||
U12983.1 | - | 3, 4 | 751 | |||
Z48781.1 | - | 3, 4 | 1214 |